Learn More
Medchemexpress LLC Gamma-glutamylcysteine | 636-58-8 | MFCD00237863 | 95.2% | 250.27 g·mol⁻¹ | C8H14N2O5S | 5 MG
SDP
Catalog No. 5000264183
Encompass_Preferred
Gamma-glutamylcysteine is a dipeptide of glutamic acid and cysteine that serves as the immediate biosynthetic precursor to glutathione. It is supplied as a white to off-white solid for research and analytical applications in studies of antioxidant pathways and glutathione metabolism.
  • Precursor to glutathione and useful for modulating intracellular GSH levels.
  • High purity (~95.2%) suitable for research and analytical use.
  • Molecular weight 250.27 g·mol⁻¹; molecular formula C8H14N2O5S.
  • White to off-white solid appearance; stable when stored under recommended conditions.
  • Available in small research pack sizes for laboratory assays.
Catalog No. 50-002-64183 Supplier Medchemexpress LLC Supplier No. HY1134025MG
Gamma-glutamylcysteine is a dipeptide of glutamic acid and cysteine that serves as the immediate biosynthetic precursor to glutathione. It is supplied as a white to off-white solid for research and analytical applications in studies of antioxidant pathways and glutathione metabolism.
  • Precursor to glutathione and useful for modulating intracellular GSH levels.
  • High purity (~95.2%) suitable for research and analytical use.
  • Molecular weight 250.27 g·mol⁻¹; molecular formula C8H14N2O5S.
  • White to off-white solid appearance; stable when stored under recommended conditions.
  • Available in small research pack sizes for laboratory assays.