Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-glutamylcysteine TFA | 283159-88-6 | 95.2% | 364.30 | 10 MG
SDP

Catalog No. 5000436646
Encompass_Preferred

Gamma-glutamylcysteine TFA is an orally active, blood-brain barrier permeable dipeptide involved in regulating macrophage polarization, inducing glutathione synthesis, and improving metabolic dysfunction. It is relevant for research in various diseases.

  • Orally active and blood-brain barrier permeable dipeptide.
  • Activates AMPK, SIRT1, IL-4/STAT6, AC/cAMP/PI3K, IGF-1R/IRS1/PI3K, and Nrf2 signaling pathways.
  • Inhibits NF-κB, JAK1/STAT1/3, MAPKs, and PI3K/Akt signaling pathways.
  • Regulates macrophage polarization and modulates CD36 and GLUT4 trafficking.
  • Induces glutathione synthesis, improves metabolic dysfunction, and ameliorates glucose homeostasis.
  • Reduces lipid deposition, inhibits apoptosis, stabilizes mitochondria, and suppresses ferroptosis.
  • Relevant for research in inflammatory bowel disease, type 2 diabetes, Alzheimer's disease, and alcoholic liver disease.

Catalog No. 50-004-36646 Supplier Medchemexpress LLC Supplier No. HY113402A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-glutamylcysteine TFA is an orally active, blood-brain barrier permeable dipeptide involved in regulating macrophage polarization, inducing glutathione synthesis, and improving metabolic dysfunction. It is relevant for research in various diseases.

  • Orally active and blood-brain barrier permeable dipeptide.
  • Activates AMPK, SIRT1, IL-4/STAT6, AC/cAMP/PI3K, IGF-1R/IRS1/PI3K, and Nrf2 signaling pathways.
  • Inhibits NF-κB, JAK1/STAT1/3, MAPKs, and PI3K/Akt signaling pathways.
  • Regulates macrophage polarization and modulates CD36 and GLUT4 trafficking.
  • Induces glutathione synthesis, improves metabolic dysfunction, and ameliorates glucose homeostasis.
  • Reduces lipid deposition, inhibits apoptosis, stabilizes mitochondria, and suppresses ferroptosis.
  • Relevant for research in inflammatory bowel disease, type 2 diabetes, Alzheimer's disease, and alcoholic liver disease.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.