Learn More
REVMAB BIOSCIENCES USA INC Gamma H2AX (phospho-Ser139) [RM224], Rabbit Monoclonal Antibody)
Supplier:  REVMAB BIOSCIENCES USA INC 31110400
Encompass_Preferred
Anti-Gamma H2AX (phospho-Ser139) Rabbit Monoclonal Antibody, Clone RM224 (100UG); WB, ELISA, Multiplex, ICC validated; Product info: www.revmab.com
Catalog No. NC1729101