Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma Hexalactone natural500G
Catalog No. 502907035
Encompass
I3-Hexalactone is one the key volatile aroma compounds of peaches. It is also found in pawpaw fruit.
Catalog No. 50-290-7035 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W255611500GK
I3-Hexalactone is one the key volatile aroma compounds of peaches. It is also found in pawpaw fruit.