Learn More
Keystone Cleanroom Products Gamma-Irradiated Fine Point Sharpie
SDP
Catalog No. 17010186
Change view
Click to view available options
Color:
Black
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-010-186 Blue
17-010-185 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-010-186 Supplier Keystone Cleanroom Products Supplier No. S01FMARKBLUE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Tip Style Fine
Product Type Cleanroom Marker
Color Blue
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation
No. per Case 500/Cs.
Length (Metric) 14 cm