Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma-L-Glutamyl-4-nitroanilide, 98+%, Thermo Scientific™

Catalog No. AAA1444214
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444214 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1444214 Supplier Thermo Scientific Chemicals Supplier No. A1444214

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7300-59-6
Molecular Formula C11H13N3O5
Quantity 25g
MDL Number MFCD00036218
Beilstein 2818758
Synonym s-2-amino-5-4-nitrophenyl amino-5-oxopentanoic acid, l-glutamine, n-4-nitrophenyl, n-4-nitrophenyl-l-glutamine, l-gamma-glutamyl-p-nitroanilide, unii-81sz84nxws, l-2-amino-4'-nitroglutaranilic acid, gamma-glutamine-4-nitroanilide, gamma-l-glutamyl-4-nitroanilide, 81sz84nxws, chembl6251
InChI Key WMZTYIRRBCGARG-VIFPVBQESA-N
SMILES [NH3+][C@@H](CCC(=O)NC1=CC=C(C=C1)[N+]([O-])=O)C([O-])=O
IUPAC Name (2S)-2-azaniumyl-4-[(4-nitrophenyl)carbamoyl]butanoate
Molecular Weight (g/mol) 267.24
PubChem CID 81732
Formula Weight 267.24
Percent Purity ≥98%
Chemical Name or Material gamma-L-Glutamyl-4-nitroanilide
Show More Show Less
EINECSNumber 230-748-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.