Learn More
TARGETMOL CHEMICALS INC GAMMA-MANGOSTIN 10MG
SDP
Catalog No. 502579830
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. gamma-Mangostin (Normangostin) as a preventive agent of the metabolic syndrome. 2. Gamma-Mangostin has free radical scavenging activity and antiproliferative and apoptotic activity in HepG2 cells. 3. Gamma-Mangostin could serve as a micronutrient for colon cancer prevention and is a potential lead compound for the development of anti-colon cancer agents. 4. Gamma-Mangostin may acts as an antihypertensive agent by causing vasorelaxation which is mediated via the NO-cGMP pathway. purity: 99%
Catalog No. 50-257-9830 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S163710MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. gamma-Mangostin (Normangostin) as a preventive agent of the metabolic syndrome. 2. Gamma-Mangostin has free radical scavenging activity and antiproliferative and apoptotic activity in HepG2 cells. 3. Gamma-Mangostin could serve as a micronutrient for colon cancer prevention and is a potential lead compound for the development of anti-colon cancer agents. 4. Gamma-Mangostin may acts as an antihypertensive agent by causing vasorelaxation which is mediated via the NO-cGMP pathway. purity: 99%