Learn More
Medchemexpress LLC Gamma-Mangostin | 31271-07-5 | 396.43 | 20 MG
SDP
Catalog No. 5000414900
Encompass_Preferred
Gamma-Mangostin is a novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist and potent epoxidase 2 (COX-2) inhibitor, as well as a transthyroxin protein (TTR) profibrosis inhibitor. It binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer. It possesses anti-inflammatory, antibacterial, antioxidant, and anticancer activities, and can be used in the study of metabolic disorders such as diabetes.
  • Novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist
  • Potent epoxidase 2 (COX-2) inhibitor
  • Transthyroxin protein (TTR) profibrosis inhibitor
  • Binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer
  • Anti-inflammatory activity
  • Antibacterial activity
  • Antioxidant activity
  • Anticancer activities
  • Used in the study of metabolic disorders such as diabetes
Catalog No. 50-004-14900 Supplier Medchemexpress LLC Supplier No. HYN195720MG
Gamma-Mangostin is a novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist and potent epoxidase 2 (COX-2) inhibitor, as well as a transthyroxin protein (TTR) profibrosis inhibitor. It binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer. It possesses anti-inflammatory, antibacterial, antioxidant, and anticancer activities, and can be used in the study of metabolic disorders such as diabetes.
  • Novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist
  • Potent epoxidase 2 (COX-2) inhibitor
  • Transthyroxin protein (TTR) profibrosis inhibitor
  • Binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer
  • Anti-inflammatory activity
  • Antibacterial activity
  • Antioxidant activity
  • Anticancer activities
  • Used in the study of metabolic disorders such as diabetes