Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-mangostin | 31271-07-5 | MFCD00951752 | 99.9% | 396.43 | C23H24O6 | 10 MG
SDP

Catalog No. 5000296392
Encompass_Preferred

Gamma-mangostin is a natural xanthone research compound with demonstrated activity as a competitive 5-HT2A receptor antagonist and a potent COX-2 inhibitor. It is used in studies of inflammation, receptor pharmacology, and transthyretin (TTR) stabilization. Supplied as a high-purity solid with recommended DMSO solubility and established in vivo formulation protocols.

  • High purity suitable for biochemical and pharmacology assays.
  • Chemical formula C23H24O6; molecular weight 396.43.
  • Reported activities include 5-HT2A antagonism, COX-2 inhibition, and TTR stabilization.
  • Available in small milligram quantities (including 10 MG) for screening and dose-response studies.
  • Soluble in DMSO with manufacturer-recommended in vivo formulation options.

Catalog No. 50-002-96392 Supplier Medchemexpress LLC Supplier No. HYN195710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-mangostin is a natural xanthone research compound with demonstrated activity as a competitive 5-HT2A receptor antagonist and a potent COX-2 inhibitor. It is used in studies of inflammation, receptor pharmacology, and transthyretin (TTR) stabilization. Supplied as a high-purity solid with recommended DMSO solubility and established in vivo formulation protocols.

  • High purity suitable for biochemical and pharmacology assays.
  • Chemical formula C23H24O6; molecular weight 396.43.
  • Reported activities include 5-HT2A antagonism, COX-2 inhibition, and TTR stabilization.
  • Available in small milligram quantities (including 10 MG) for screening and dose-response studies.
  • Soluble in DMSO with manufacturer-recommended in vivo formulation options.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.