Learn More
CPC Scientific H-Tyr-Val-Met-Gly-His-Phe-Arg-Trp-Asp-Arg-Phe-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MSHS007B
Encompass
Fragment that corresponds to cryptic region of the ACTH/beta-lipotropin precursor protein from the intermediate lobe of the bovine pituitary. ONE-LETTER SEQUENCE: YVMGHFRWDRFGMOLECULAR FORMULA: C74H99N21O16S1MOLECULAR WEIGHT:1570.77STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [72629-65-3], SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES: S. Nakanishi et al., Nature, 278, 423 (1979)
Catalog No. 50-210-9846