Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich gamma-Octanoic lactone

Catalog No. O400825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O400825G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O400825G Supplier Sigma Aldrich Supplier No. O400825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl 2,6-dibromoheptanedioate acts as a bifunctional initiator in various polymerization reactions. its electrochemical cyclization affords high yields of three- to six-membered dimethyl 1,2-cycloalkanedicarboxylates.dimethyl 2,6-dibromoheptanedioate may be used in the following studies:
  • Preparation of difunctional poly(n-butyl acrylate) (pba) macroinitiator
  • As initiator for the synthesis of dibromo-terminated polystyrene, via atom transfer radical polymerization (atrp)
  • Preparation of polytrithiocarbonate, which serves as reversible addition-fragmentation chain transfer (raft) agent for radical polymerization reactions

Specifications

CAS 104-50-7
Density 0.981 g/mL (at 25°C (literature))
Boiling Point 234°C (lit.)
Molecular Formula C8H14O2
Refractive Index n20/D 1.444 (literature)
Linear Formula C8H14O2
MDL Number MFCD00005402
Quantity 10 mL
Synonym 4-Hydroxy octanoic acid; Octano-1,4-lactone
Molecular Weight (g/mol) 142.2
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.