Learn More
Selleck Chemical LLC Gamma-Oryzanol S3957-25mg
SDP
Catalog No. 503108113
Encompass_Preferred
Gamma-oryzanol (GO) a nutriactive phytochemical naturally occurring in crude rice bran oil is an antioxidant compound used to treat hyperlipidemia disorders of menopause and to increase the muscle mass -Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice -Oryzanol significantly inhibits the activities of DNMT1 (IC50 3 2 M) DNMT3a (IC50 22 3 M)
Catalog No. 50-310-8113 Supplier Selleck Chemical LLC Supplier No. S395725MG
Gamma-oryzanol (GO) a nutriactive phytochemical naturally occurring in crude rice bran oil is an antioxidant compound used to treat hyperlipidemia disorders of menopause and to increase the muscle mass -Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice -Oryzanol significantly inhibits the activities of DNMT1 (IC50 3 2 M) DNMT3a (IC50 22 3 M)