Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma-Oryzanol, TCI America™
SDP

Catalog No. O0172250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0172250G 250 g
O017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0172250G Supplier TCI America Supplier No. O0172250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 11042-64-1
Molecular Formula C40H58O4
Molecular Weight (g/mol) 602.90
MDL Number MFCD00867548
InChI Key FODTZLFLDFKIQH-FSVGXZBPSA-N
PubChem CID 91971120
IUPAC Name (1S,3R,6S,8R,11S,12S,15R,16R)-7,7,12,16-tetramethyl-15-[(2R)-6-methylhept-5-en-2-yl]pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-6-yl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES COC1=CC(\C=C\C(=O)O[C@H]2CC[C@]34C[C@]33CC[C@]5(C)[C@H](CC[C@@]5(C)[C@@H]3CC[C@H]4C2(C)C)[C@H](C)CCC=C(C)C)=CC=C1O

Specifications

Color White-Yellow
Quantity 250 g
UN Number 2811
Physical Form Crystalline Powder
Chemical Name or Material gamma-Oryzanol
EINECSNumber (8)-0485
RTECSNumber RN0050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.