Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gamma-[(Propargyl)-Imido]-Atp Sodium Salt

Catalog No. 8085121MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-851-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-851-21MG Supplier Sigma Aldrich Supplier No. 8085121MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lee, et al,. reported a non-radioactive version of in vitro phosphorylation where +a-[(Propargyl)-imido]-ATP (compound 1) was successfully used instead of +a-32P-modified ATP to phosphorylate GST-tagged recombinant p27kip1 with protein kinase cdk2. The phosphorylated, alkyne-modified protein substrate can be labeled with azides of biotin or fluorescent dyes using Cu(I)-catalyzed click chemistry.The purchase of this product for use in applications relating to copper catalyzed azide-alkyne cycloaddition chemistry (Click Chemistry) includes a limited, nontransferable license to intellectual property owned by The Scripps Research Institute to use this product solely for internal non-commercial research activities and specifically excludes clinical, therapeutic, or diagnostic use in humans or animals and manufacturing purposes. Information regarding a license for commercial use in Click Chemistry may be obtained directly from The Scripps Research Institute, 10550 N. Torrey Pines Rd., La Jolla, CA 92037, or by contacting 858-784-8140 or click@scripps.edu.

Specifications

CAS 207386-83-2
Molecular Formula C16H28N9O12P3 -+ xNa+
Linear Formula C16H28N9O12P3 -+ xNa+
Quantity 1 mg
Synonym gamma-[(6-Azidohexyl)-imido]-adenosine-5'-triphosphate sodium salt
Molecular Weight (g/mol) 631.37 (free acid basis) g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.