Learn More
AdipoGen gamma-Rubromycin
SDP
Catalog No. 502562152
Encompass_Preferred
Chemical. CAS 27267-71-6. Formula C26H18O12. MW 522.4. Isolated from Streptomyces sp. Antibiotic. HIV-1 reverse transcriptase and human telomerase inhibitor. Antibacterial. Cytostatically active against different tumor cell lines.
Catalog No. 50-256-2152 Supplier AdipoGen Supplier No. BVT0007M005
Add to Cart
Add to Cart
Chemical. CAS 27267-71-6. Formula C26H18O12. MW 522.4. Isolated from Streptomyces sp. Antibiotic. HIV-1 reverse transcriptase and human telomerase inhibitor. Antibacterial. Cytostatically active against different tumor cell lines.