Learn More
Uline GAMMA SEAL LID RED
Catalog No. NC2026608
Encompass_Preferred
Screw-on lid for 5-gallon buckets creates an airtight seal to protect contents with a rotating center for access without unscrewing the entire lid; made from FDA approved, food grade materials in red color.
Catalog No. NC2026608 Supplier Uline Supplier No. S17945R
Screw-on lid for 5-gallon buckets creates an airtight seal to protect contents with a rotating center for access without unscrewing the entire lid; made from FDA approved, food grade materials in red color.