Learn More
US Plastic Corporation GAMMA SEALS SCREW LIDS-RED
SDP
Catalog No. NC0204598
Encompass
Red Gamma Seals™ Turn pails into reusable containers. Heavy duty, high density polyethylene construction creates a leak-proof airtight seal. Resin meets FDA standards. Simply tap on the adapter ring (includes rubber gasket) and screw inner cover on
This item is not returnable. View return policy
Catalog No. NC0204598 Supplier US Plastic Corporation Supplier No. 1861
Red Gamma Seals™ Turn pails into reusable containers. Heavy duty, high density polyethylene construction creates a leak-proof airtight seal. Resin meets FDA standards. Simply tap on the adapter ring (includes rubber gasket) and screw inner cover on