Learn More
Cell Signaling Technology Gamma Secretase Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502732755
Encompass
The γ Secretase Antibody Sampler Kit provides an economical means of evaluating components of the gamma secretase complex. This kit contains primary and secondary antibodies sufficient to perform two western miniblot experiments.
  • Includes antibodies for PEN2, presenilin-2, nicastrin, and presenilin-1
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Optimized for western blotting and immunoprecipitation
  • Convenient sampler format for efficient protein complex analysis
Catalog No. 50-273-2755 Supplier Cell Signaling Technology Supplier No. 5887T
The γ Secretase Antibody Sampler Kit provides an economical means of evaluating components of the gamma secretase complex. This kit contains primary and secondary antibodies sufficient to perform two western miniblot experiments.
  • Includes antibodies for PEN2, presenilin-2, nicastrin, and presenilin-1
  • Supplied with HRP-linked anti-rabbit IgG secondary antibody
  • Optimized for western blotting and immunoprecipitation
  • Convenient sampler format for efficient protein complex analysis