Learn More
Medchemexpress LLC Gamma-secretase modulator 1 | 1172637-87-4 | 98.0% | 416.54 | C24H24N4OS | 100 MG
SDP
Catalog No. 5000214819
Encompass_Preferred
Gamma-secretase modulator 1 is a research-grade small molecule that modulates γ-secretase activity and is used to study amyloid-β production and Alzheimer's disease mechanisms. It is supplied as a high-purity solid suitable for in vitro and mechanistic research.
  • High purity (98.0%) for reproducible experimental results.
  • Characterized molecular properties: C24H24N4OS, MW 416.54.
  • Suitable for in vitro and mechanistic studies of γ-secretase and amyloid-β production.
  • Supplied as a solid with multiple pack size options.
  • Stable when stored under recommended conditions for research reagents.
Catalog No. 50-002-14819 Supplier Medchemexpress LLC Supplier No. HY10043100MG
Gamma-secretase modulator 1 is a research-grade small molecule that modulates γ-secretase activity and is used to study amyloid-β production and Alzheimer's disease mechanisms. It is supplied as a high-purity solid suitable for in vitro and mechanistic research.
  • High purity (98.0%) for reproducible experimental results.
  • Characterized molecular properties: C24H24N4OS, MW 416.54.
  • Suitable for in vitro and mechanistic studies of γ-secretase and amyloid-β production.
  • Supplied as a solid with multiple pack size options.
  • Stable when stored under recommended conditions for research reagents.