Learn More
Medchemexpress LLC Gamma-secretase modulator 1 | 1172637-87-4 | 98.0% | 416.54 g/mol | C24H24N4OS | 50 MG
SDP
Catalog No. 5000215006
Encompass_Preferred
Gamma-secretase modulator 1 is a small-molecule research reagent that modulates γ-secretase activity to alter amyloid-β peptide production. It is supplied as a solid for preclinical laboratory studies and intended for research use only; not for diagnostic or therapeutic use.
  • Modulates γ-secretase activity to reduce Aβ42 production.
  • Suitable for biochemical and cellular assays.
  • High purity (98.0%) for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 416.54 g/mol; formula C24H24N4OS.
  • Powder stable at -20°C for long-term storage, 4°C for short-term storage.
  • Supplied in a 50 MG quantity for small-scale studies.
  • For research use only; not for human administration.
Catalog No. 50-002-15006 Supplier Medchemexpress LLC Supplier No. HY1004350MG
Gamma-secretase modulator 1 is a small-molecule research reagent that modulates γ-secretase activity to alter amyloid-β peptide production. It is supplied as a solid for preclinical laboratory studies and intended for research use only; not for diagnostic or therapeutic use.
  • Modulates γ-secretase activity to reduce Aβ42 production.
  • Suitable for biochemical and cellular assays.
  • High purity (98.0%) for reproducible results.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 416.54 g/mol; formula C24H24N4OS.
  • Powder stable at -20°C for long-term storage, 4°C for short-term storage.
  • Supplied in a 50 MG quantity for small-scale studies.
  • For research use only; not for human administration.