Learn More
Medchemexpress LLC Gamma-secretase modulator 1 hydrochloride | 2741571-83-3 | 99.9% | 453.00 | C24H25ClN4OS | 100 MG
SDP
Catalog No. 5000215067
Encompass_Preferred
Gamma-secretase modulator 1 hydrochloride is a small-molecule gamma-secretase modulator supplied as the hydrochloride salt for laboratory research. It is described by the manufacturer as useful for Alzheimer's disease research and is provided with analytical characterization for research use only.
  • Modulates γ-secretase activity in biochemical and cellular assays.
  • Supplied as hydrochloride salt to aid solubility in assays.
  • High purity suitable for research (>99.8%).
  • Recommended storage: 4°C sealed; in solvent -80°C for 6 months or -20°C for 1 month.
  • Not validated for medical or clinical use; for research use only.
Catalog No. 50-002-15067 Supplier Medchemexpress LLC Supplier No. HY10043A100MG
Gamma-secretase modulator 1 hydrochloride is a small-molecule gamma-secretase modulator supplied as the hydrochloride salt for laboratory research. It is described by the manufacturer as useful for Alzheimer's disease research and is provided with analytical characterization for research use only.
  • Modulates γ-secretase activity in biochemical and cellular assays.
  • Supplied as hydrochloride salt to aid solubility in assays.
  • High purity suitable for research (>99.8%).
  • Recommended storage: 4°C sealed; in solvent -80°C for 6 months or -20°C for 1 month.
  • Not validated for medical or clinical use; for research use only.