Learn More
Medchemexpress LLC Gamma-secretase modulator 1 hydrochloride | 2741571-83-3 | 99.9% | 453.00 g·mol⁻¹ | C24H25ClN4OS | 1 ML
SDP
Catalog No. 5000214563
Encompass_Preferred
Gamma-secretase modulator 1 (hydrochloride) is a small-molecule γ-secretase modulator used in preclinical research to study amyloid precursor protein processing and mechanisms related to Alzheimer's disease. The product is available as a high-purity powder or as a prepared 10 mM solution in DMSO (1 mL), providing a convenient, ready-to-use option for biochemical and cellular assays. Research use only; follow supplier guidance for storage and handling.
  • High purity suitable for analytical and biological studies.
  • Hydrochloride salt with defined molecular formula and molecular weight.
  • Available as 10 mM solution in DMSO (1 mL) and as powder formats.
  • Useful for studies of amyloid precursor protein processing and Alzheimer's disease mechanisms.
  • Store powder at 4°C and protect from moisture; handle solutions per supplier guidance.
Catalog No. 50-002-14563 Supplier Medchemexpress LLC Supplier No. HY10043A10MM 1ML
Gamma-secretase modulator 1 (hydrochloride) is a small-molecule γ-secretase modulator used in preclinical research to study amyloid precursor protein processing and mechanisms related to Alzheimer's disease. The product is available as a high-purity powder or as a prepared 10 mM solution in DMSO (1 mL), providing a convenient, ready-to-use option for biochemical and cellular assays. Research use only; follow supplier guidance for storage and handling.
  • High purity suitable for analytical and biological studies.
  • Hydrochloride salt with defined molecular formula and molecular weight.
  • Available as 10 mM solution in DMSO (1 mL) and as powder formats.
  • Useful for studies of amyloid precursor protein processing and Alzheimer's disease mechanisms.
  • Store powder at 4°C and protect from moisture; handle solutions per supplier guidance.