Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-secretase modulator 2 | 1093978-89-2 | 98.8% | C25H22F4N6O2 | 1 ML
SDP

Catalog No. 5000358083
Encompass_Preferred

Gamma-secretase modulator 2 is a potent and selective gamma-secretase modulator. It is used for the study of Alzheimer's disease and is intended for research use only.

  • Potent and selective gamma-secretase modulator
  • Appears as a solid
  • Color is white to off-white
  • Soluble in DMSO at 100 mg/mL
  • Useful for research in neurological and neurodegenerative diseases

Catalog No. 50-003-58083 Supplier Medchemexpress LLC Supplier No. HY5075410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-secretase modulator 2 is a potent and selective gamma-secretase modulator. It is used for the study of Alzheimer's disease and is intended for research use only.

  • Potent and selective gamma-secretase modulator
  • Appears as a solid
  • Color is white to off-white
  • Soluble in DMSO at 100 mg/mL
  • Useful for research in neurological and neurodegenerative diseases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.