Learn More
Medchemexpress LLC Gamma-secretase modulator 2 | 1093978-89-2 | 98.80% | C25H22F4N6O2 | 5 MG
SDP
Catalog No. 5000358191
Encompass_Preferred
Gamma-secretase modulator 2 is a potent and selective γ-secretase modulator, intended for the study of Alzheimer's disease. It is for research use only and not for sale to patients. The compound presents as a white to off-white solid with a molecular weight of 514.47.
  • Potent and selective γ-secretase modulator
  • Used for the study of Alzheimer's disease
  • White to off-white solid appearance
Catalog No. 50-003-58191 Supplier Medchemexpress LLC Supplier No. HY507545MG
Gamma-secretase modulator 2 is a potent and selective γ-secretase modulator, intended for the study of Alzheimer's disease. It is for research use only and not for sale to patients. The compound presents as a white to off-white solid with a molecular weight of 514.47.
  • Potent and selective γ-secretase modulator
  • Used for the study of Alzheimer's disease
  • White to off-white solid appearance