Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-secretase modulator 3 | 1431697-84-5 | 98.0% | C24H23FN4OS | 100 MG
SDP

Catalog No. 5000357705
Encompass_Preferred

gamma-secretase modulator 3 is a gamma-secretase modulator intended for research use only.

  • Molecular weight: 434.53
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping conditions: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): DMSO: < 1 mg/mL (insoluble or slightly soluble)
  • For research use only, not for sale to patients

Catalog No. 50-003-57705 Supplier Medchemexpress LLC Supplier No. HY50889100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

gamma-secretase modulator 3 is a gamma-secretase modulator intended for research use only.

  • Molecular weight: 434.53
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping conditions: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): DMSO: < 1 mg/mL (insoluble or slightly soluble)
  • For research use only, not for sale to patients

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.