Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-secretase modulator 3 | 1431697-84-5 | 98.0% | C24H23FN4OS | 50 MG
SDP

Supplier:  Medchemexpress LLC HY5088950MG

Encompass_Preferred

Gamma-secretase modulator 3 is a compound identified as a gamma-secretase modulator, meaning it interacts with and modulates the activity of the gamma-secretase enzyme. This enzyme is a multi-protein complex known for its role in the intramembrane cleavage of various proteins, including Amyloid Precursor Protein (APP), which is implicated in Alzheimer's disease pathogenesis.

  • Functions as a gamma-secretase modulator.
  • Appearance: solid, light yellow to yellow.
  • Molecular weight: 434.53.
  • Intended for research purposes only.

Catalog No. 50-003-57801


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.