Learn More
Medchemexpress LLC Gamma-secretase modulator 6 | 1255700-88-9 | 99.7% | 10 MG
SDP
Catalog No. 5000295415
Encompass_Preferred
Gamma-secretase modulator 6 is a high-purity research compound that modulates gamma-secretase activity and reduces Aβ42 secretion in cellular models (pIC50 8.1). It is provided as a solid or as a DMSO solution for preclinical biochemical and cellular research into Alzheimer's disease mechanisms.
  • Inhibits Aβ42 secretion (pIC50 8.1).
  • High purity suitable for research (99.68%).
  • Available as solid and as a 10 mM DMSO solution.
  • Molecular formula C25H26N6O2, molecular weight 442.51.
  • Intended for preclinical and biochemical studies of amyloid processing.
Catalog No. 50-002-95415 Supplier Medchemexpress LLC Supplier No. HY17134710MG
Gamma-secretase modulator 6 is a high-purity research compound that modulates gamma-secretase activity and reduces Aβ42 secretion in cellular models (pIC50 8.1). It is provided as a solid or as a DMSO solution for preclinical biochemical and cellular research into Alzheimer's disease mechanisms.
  • Inhibits Aβ42 secretion (pIC50 8.1).
  • High purity suitable for research (99.68%).
  • Available as solid and as a 10 mM DMSO solution.
  • Molecular formula C25H26N6O2, molecular weight 442.51.
  • Intended for preclinical and biochemical studies of amyloid processing.