Learn More
Medchemexpress LLC Gamma-secretase modulator 6 | 1255700-88-9 | 99.7% | 442.51 | 1 ML
SDP
Catalog No. 5000386184
Encompass_Preferred
gamma-secretase modulator 6 (Example 50) is a gamma-secretase modulator. It inhibits Aβ42 secretion in HEK cell line stably expressing APP (Aβ amyloid precursor protein) (pIC50: 8.1), making it suitable for research.
  • Used for research of Alzheimer's disease
  • Inhibits Aβ42 secretion in HEK cell lines stably expressing APP
Catalog No. 50-003-86184 Supplier Medchemexpress LLC Supplier No. HY17134710MM/1ML
gamma-secretase modulator 6 (Example 50) is a gamma-secretase modulator. It inhibits Aβ42 secretion in HEK cell line stably expressing APP (Aβ amyloid precursor protein) (pIC50: 8.1), making it suitable for research.
  • Used for research of Alzheimer's disease
  • Inhibits Aβ42 secretion in HEK cell lines stably expressing APP