Learn More
Medchemexpress LLC gamma-Secretase Modulators | 937812-80-1 | 99.8% | C26H24F3N3O3 | 1 MG
SDP
Catalog No. 5000357747
Encompass_Preferred
This product, also known as Amyloid-β production inhibitor, is useful for Alzheimer's disease research. It is a solid appearing white to off-white and acts as a γ-secretase modulator.
  • Useful for Alzheimer's disease research
  • Targets γ-secretase modulator
  • Solid with a white to off-white appearance
  • Shipped at room temperature in continental US
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-003-57747 Supplier Medchemexpress LLC Supplier No. HY509001MG
This product, also known as Amyloid-β production inhibitor, is useful for Alzheimer's disease research. It is a solid appearing white to off-white and acts as a γ-secretase modulator.
  • Useful for Alzheimer's disease research
  • Targets γ-secretase modulator
  • Solid with a white to off-white appearance
  • Shipped at room temperature in continental US
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen