Learn More
CROSSTEX INTERNATIONAL GAMMA STERLZATION PLANE 5000RL
Catalog No. NC0013935
Encompass_Preferred
Radiation Chemical Process Indicators, 1/2 (12.7mm) 5000 Indicators/roll, ISO 11140-1 Class 1 compliant. Yellow to Red color change.
Catalog No. NC0013935 Supplier CROSSTEX INTERNATIONAL Supplier No. NMCPIR01
Radiation Chemical Process Indicators, 1/2 (12.7mm) 5000 Indicators/roll, ISO 11140-1 Class 1 compliant. Yellow to Red color change.