Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-terpinene | 99-85-4 | MFCD00001537 | 98.1% | 136.23 g·mol⁻¹ | C10H16 | 1 ML
SDP

Catalog No. 5000301980
Encompass_Preferred

Gamma-terpinene is a monoterpene antioxidant commonly used as an analytical reference standard and reagent in biochemical and pharmacological research. This product is supplied as a ready-to-use 10 mM solution in DMSO, offering convenience for assays and stock preparation while meeting reported purity specifications.

  • Monoterpene antioxidant used as a reference standard.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • High purity, 98.14% (manufacturer reported).
  • Molecular weight 136.23 g·mol⁻¹; formula C10H16.
  • CAS number 99-85-4 for unambiguous identification.
  • Suitable for analytical, biochemical, and pharmacology studies.

Catalog No. 50-003-01980 Supplier Medchemexpress LLC Supplier No. HYW02018310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-terpinene is a monoterpene antioxidant commonly used as an analytical reference standard and reagent in biochemical and pharmacological research. This product is supplied as a ready-to-use 10 mM solution in DMSO, offering convenience for assays and stock preparation while meeting reported purity specifications.

  • Monoterpene antioxidant used as a reference standard.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • High purity, 98.14% (manufacturer reported).
  • Molecular weight 136.23 g·mol⁻¹; formula C10H16.
  • CAS number 99-85-4 for unambiguous identification.
  • Suitable for analytical, biochemical, and pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.