Learn More
TARGETMOL CHEMICALS INC GAMMA-TOCOTRIENOL 10MG
SDP
Catalog No. 502581044
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (gamma)-Tocotrienol (Plastochromanol) is one of the four types of tocotrienol a type of vitamin E. Itl is also a radioprotector antioxidant. It shows antitumor and antihypertensive effects in vivo. purity: 98%
Catalog No. 50-258-1044 Supplier TARGETMOL CHEMICALS INC Supplier No. T1976810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (gamma)-Tocotrienol (Plastochromanol) is one of the four types of tocotrienol a type of vitamin E. Itl is also a radioprotector antioxidant. It shows antitumor and antihypertensive effects in vivo. purity: 98%