Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma-undecalactone | 104-67-6 | MFCD00005405 | 99.0% | 184.27 g/mol | C11H20O2 | 100 MG
SDP

Catalog No. 5000302228
Encompass_Preferred

Gamma-undecalactone is a naturally occurring lactone with a characteristic fruity, peach-like aroma used as a flavor and fragrance ingredient and as a biochemical reagent in research. It is supplied as a high-purity liquid suitable for analytical, formulation, and assay applications.

  • Molecular formula: C11H20O2.
  • Molecular weight: 184.27 g/mol.
  • CAS number: 104-67-6.
  • Physical form: colorless to light yellow liquid.
  • Purity: ~99% (high purity).
  • Typical uses: flavoring, fragrance, biochemical research.
  • Solubility: soluble in DMSO.

Catalog No. 50-003-02228 Supplier Medchemexpress LLC Supplier No. HYW016278100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-undecalactone is a naturally occurring lactone with a characteristic fruity, peach-like aroma used as a flavor and fragrance ingredient and as a biochemical reagent in research. It is supplied as a high-purity liquid suitable for analytical, formulation, and assay applications.

  • Molecular formula: C11H20O2.
  • Molecular weight: 184.27 g/mol.
  • CAS number: 104-67-6.
  • Physical form: colorless to light yellow liquid.
  • Purity: ~99% (high purity).
  • Typical uses: flavoring, fragrance, biochemical research.
  • Solubility: soluble in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.