Learn More
Medchemexpress LLC Gamma-undecalactone | 104-67-6 | MFCD00005405 | 99.0% | 184.28 | C11H20O2 | 50 MG
SDP
Catalog No. 5000302226
Encompass_Preferred
Gamma-undecalactone (CAS 104-67-6) is a gamma-lactone compound used as a biochemical reagent and as a flavor and fragrance intermediate in research and analytical settings. It serves as a chromatographic standard and sensory reference material for formulation, analytical method development, and flavor chemistry studies.
  • Used as a biochemical reagent and flavor/fragrance intermediate.
  • High purity: 98.96% as stated by the supplier.
  • Molecular weight 184.28 g/mol for stoichiometric and analytical calculations.
  • Supplied in small analytical quantities suitable for method development and standards.
  • Compatible with common organic solvents such as DMSO for sample preparation.
Catalog No. 50-003-02226 Supplier Medchemexpress LLC Supplier No. HYW01627850MG
Gamma-undecalactone (CAS 104-67-6) is a gamma-lactone compound used as a biochemical reagent and as a flavor and fragrance intermediate in research and analytical settings. It serves as a chromatographic standard and sensory reference material for formulation, analytical method development, and flavor chemistry studies.
  • Used as a biochemical reagent and flavor/fragrance intermediate.
  • High purity: 98.96% as stated by the supplier.
  • Molecular weight 184.28 g/mol for stoichiometric and analytical calculations.
  • Supplied in small analytical quantities suitable for method development and standards.
  • Compatible with common organic solvents such as DMSO for sample preparation.