Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma Valerolactone BioRen
Catalog No. 502903629
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. gamma-ValerolactoneA(GVL) is a bio-based fully degradable non-toxic green solvent and has been aligned with Safer Solvent and Auxiliaries Use of Renewable feedstocks and Design for Degradation. Click here for more information.
Catalog No. 50-290-3629 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9186604L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. gamma-ValerolactoneA(GVL) is a bio-based fully degradable non-toxic green solvent and has been aligned with Safer Solvent and Auxiliaries Use of Renewable feedstocks and Design for Degradation. Click here for more information.