Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma-Valerolactone BioRen1L
Catalog No. 502897533
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. gamma-ValerolactoneA(GVL) is a bio-based fully degradable non-toxic green solvent and has been aligned with Safer Solvent and Auxiliaries Use of Renewable feedstocks and Design for Degradation. Click here for more information.
Catalog No. 50-289-7533 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9186601L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. gamma-ValerolactoneA(GVL) is a bio-based fully degradable non-toxic green solvent and has been aligned with Safer Solvent and Auxiliaries Use of Renewable feedstocks and Design for Degradation. Click here for more information.