Learn More
Sigma Aldrich Fine Chemicals Biosciences Gammabind Plus SepharoseR
Catalog No. 502907580
Encompass
GammaBind Plus Sepharose(R) is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to GammaBind Sepharose.GammaBind Plus Sepharose(R) is GammaBind G Type 3 covalently immobilized to Sepharose(R) CL-6B by malimide linkage. GammaBind G Type 3 a recombinant form of streptococcal protein G binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose(R) may be used to analyze and purify classes subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding the Fab region is still available for binding antigen.
Catalog No. 50-290-7580 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17088601
GammaBind Plus Sepharose(R) is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to GammaBind Sepharose.GammaBind Plus Sepharose(R) is GammaBind G Type 3 covalently immobilized to Sepharose(R) CL-6B by malimide linkage. GammaBind G Type 3 a recombinant form of streptococcal protein G binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose(R) may be used to analyze and purify classes subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding the Fab region is still available for binding antigen.