Learn More
Aviva Systems Biology Corp GAMT Antibody - middle region (ARP42700_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42700_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GAMT. Alternate names: PIG2, TP53I2ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-58312