Learn More
Aviva Systems Biology Corp GAMT Antibody - N-terminal region (ARP41684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GAMT. Alternate names: PIG2, TP53I2ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-57951