Learn More
Aviva Systems Biology Corp GANAB Antibody - middle region (ARP55868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GANAB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GANAB. Alternate names: G2AN, GLUII, KIAA0088ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-090