Learn More
Medchemexpress LLC HY-108024A 5mg Medchemexpress, Ganaplacide (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108024A5MG
Encompass_Preferred
Medchemexpress, HY-108024A 5mg Ganaplacide (hydrochloride) CAS:0 Ganaplacide (KAF156) hydrochloride is a first-in-class, orally active imidazolopiperazine antimalarial agent. Ganaplacide hydrochloride is active against a broad range of Plasmodium species, including drug-resistant parasites. Ganaplacide hydrochloride is parasiticidal against both asexual and sexual blood stages as well as the liver stages of the parasite[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9617