Learn More
Medchemexpress LLC Ganaplacide hydrochloride | 2751586-94-2 | 98.2% | 447.91 g/mol | C22H24ClF2N5O | 25 MG
SDP
Catalog No. 5000243019
Encompass_Preferred
Ganaplacide hydrochloride is the hydrochloride salt of ganaplacide (KAF156), an investigational antimalarial research compound supplied as a white to yellow solid for laboratory use. It has molecular formula C22H24ClF2N5O, molecular weight 447.91 g/mol, CAS 2751586-94-2, and a reported purity of 98.22%.
  • High purity suitable for research applications.
  • Hydrochloride salt form, solid and stable for handling and storage.
  • Suitable for in vitro and in vivo antimalarial pharmacology studies.
  • Characterized molecular weight and formula for analytical work.
  • Available in small research packaging for dosing flexibility.
Catalog No. 50-002-43019 Supplier Medchemexpress LLC Supplier No. HY108024A25MG
Ganaplacide hydrochloride is the hydrochloride salt of ganaplacide (KAF156), an investigational antimalarial research compound supplied as a white to yellow solid for laboratory use. It has molecular formula C22H24ClF2N5O, molecular weight 447.91 g/mol, CAS 2751586-94-2, and a reported purity of 98.22%.
  • High purity suitable for research applications.
  • Hydrochloride salt form, solid and stable for handling and storage.
  • Suitable for in vitro and in vivo antimalarial pharmacology studies.
  • Characterized molecular weight and formula for analytical work.
  • Available in small research packaging for dosing flexibility.