Learn More
Selleck Chemical LLC Ganciclovir S1878-1g
SDP
Catalog No. 503109364
Encompass_Preferred
Ganciclovir is an antiviral drug for feline herpesvirus type-1 with IC50 of 5 2 M in a cell-free assay
Catalog No. 50-310-9364 Supplier Selleck Chemical LLC Supplier No. S18781G
Ganciclovir is an antiviral drug for feline herpesvirus type-1 with IC50 of 5 2 M in a cell-free assay