Learn More
Medchemexpress LLC Ganciclovir | 82410-32-0 | 99.9% | 1 G
SDP
Catalog No. 5000349134
Encompass_Preferred
Ganciclovir (BW 759) is a nucleoside analogue and an orally active antiviral agent effective against CMV. It also exhibits in vitro activity against members of the herpes group and some other DNA viruses, including human herpes viruses (HSV 1 and 2, CMV) and adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22, and 28. It has an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1) and can diffuse into the brain.
  • Nucleoside analogue
  • Orally active antiviral agent
  • Effective against CMV, HSV, and some other DNA viruses
  • Inhibits in vitro replication of human herpes viruses (HSV 1 and 2, CMV)
  • Inhibits in vitro replication of adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22, and 28
  • IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1)
  • Can diffuse into the brain
Catalog No. 50-003-49134 Supplier Medchemexpress LLC Supplier No. HY136371G
Ganciclovir (BW 759) is a nucleoside analogue and an orally active antiviral agent effective against CMV. It also exhibits in vitro activity against members of the herpes group and some other DNA viruses, including human herpes viruses (HSV 1 and 2, CMV) and adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22, and 28. It has an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1) and can diffuse into the brain.
  • Nucleoside analogue
  • Orally active antiviral agent
  • Effective against CMV, HSV, and some other DNA viruses
  • Inhibits in vitro replication of human herpes viruses (HSV 1 and 2, CMV)
  • Inhibits in vitro replication of adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22, and 28
  • IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1)
  • Can diffuse into the brain