Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ganciclovir, 98%, Thermo Scientific Chemicals

Catalog No. AC461710010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461710010 1 g
AC461710050 5 g
AC461710250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC461710010 Supplier Thermo Scientific Chemicals Supplier No. 461710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82410-32-0
Molecular Formula C9H13N5O4
Molecular Weight (g/mol) 255.23
InChI Key IRSCQMHQWWYFCW-UHFFFAOYSA-N
Synonym ganciclovir, gancyclovir, cytovene, vitrasert, hydroxyacyclovir, ganciclovirum, zirgan, ganciclovir sodium, cymevene, hhemg
PubChem CID 3454
ChEBI CHEBI:465284
IUPAC Name 2-amino-9-(1,3-dihydroxypropan-2-yloxymethyl)-3H-purin-6-one
SMILES C1=NC2=C(N1COC(CO)CO)NC(=NC2=O)N

Specifications

Melting Point 251.0°C to 252.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 3,14g/L (25°C).
Formula Weight 255.23
Percent Purity 98%
Chemical Name or Material Ganciclovir
Hazard Category Effects on or via lactation, Germ cell mutagenicity (category 1B), Reproductive toxicity (category 1B), Carcinogenicity (category 2)
Hazard Statement May cause harm to breast-fed children, May cause genetic defects, May damage the unborn child. Suspected of damaging fertility, Suspected of causing cancer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.