Learn More
Medchemexpress LLC Ganciclovir (sodium) | 107910-75-8 | 99.9% | 277.21 | 1 ML
SDP
Catalog No. 5000348986
Encompass_Preferred
Ganciclovir (sodium) is a nucleoside analogue and an orally active antiviral agent effective against CMV. It exhibits in vitro activity against human herpes viruses (HSV 1 and 2, CMV) and various other DNA viruses, including several adenovirus serotypes. With an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1), this compound can also diffuse into the brain, making it a versatile tool for antiviral research.
  • Nucleoside analogue antiviral agent
  • Orally active and effective against CMV
  • Active against herpes group and other DNA viruses
  • Inhibits replication of human herpes viruses and adenovirus serotypes
  • IC50 of 5.2 μM for feline herpesvirus type-1
  • Capable of diffusing into the brain
  • Superior activity against CMV compared to acyclovir
  • Inhibits viral DNA replication through ganciclovir-5'-triphosphate
Catalog No. 50-003-48986 Supplier Medchemexpress LLC Supplier No. HY13637A10MM/1ML
Ganciclovir (sodium) is a nucleoside analogue and an orally active antiviral agent effective against CMV. It exhibits in vitro activity against human herpes viruses (HSV 1 and 2, CMV) and various other DNA viruses, including several adenovirus serotypes. With an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1), this compound can also diffuse into the brain, making it a versatile tool for antiviral research.
  • Nucleoside analogue antiviral agent
  • Orally active and effective against CMV
  • Active against herpes group and other DNA viruses
  • Inhibits replication of human herpes viruses and adenovirus serotypes
  • IC50 of 5.2 μM for feline herpesvirus type-1
  • Capable of diffusing into the brain
  • Superior activity against CMV compared to acyclovir
  • Inhibits viral DNA replication through ganciclovir-5'-triphosphate