Learn More
Selleck Chemical LLC Ganciclovir sodium S5065-1g
SDP
Catalog No. 503112377
Encompass_Preferred
Ganciclovir Sodium (RS-21592 Cytovene IV BW 759 2 -Nor-2 -deoxyguanosine) is the sodium salt form of ganciclovir a synthetic antiviral purine nucleoside analog with antiviral activity especially against cytomegalovirus (CMV)
Catalog No. 50-311-2377 Supplier Selleck Chemical LLC Supplier No. S50651G
Ganciclovir Sodium (RS-21592 Cytovene IV BW 759 2 -Nor-2 -deoxyguanosine) is the sodium salt form of ganciclovir a synthetic antiviral purine nucleoside analog with antiviral activity especially against cytomegalovirus (CMV)