Learn More
TOKU-E Novobiocin ReadyMade™ Solution | Novobiocin Sodium | CAS 1476-53-5 | 100 mg/ML in water | sterile-filtered | 1 ML
Supplier:  TOKU-E G09310X1ML
Encompass_Preferred
Novobiocin ReadyMade™ Solution | Novobiocin Sodium | CAS 1476-53-5 | 100 mg/ML in water | sterile-filtered | 1 ML
Catalog No. NC2068746