Learn More
Invivogen Ganciclovir (GCV), 82410-32-0, MFCD00870588, 250mg
SDP
Catalog No. NC9866266
Encompass
Molecular Formula C9H13N5O4, Molecular weight 255.2, Ganciclovir (GCV) is a guanosine analog used as a prodrug to obtain a suicide effect in cells transfected with the herpes virus thymidine kinase gene (HSV-tk). HSVTK phosphorylates GCV to GCV-monophosphate which is further converted to GCV-diphosphate and GCV-triphosphate by host kinases. GCV-triphosphate causes premature DNA chain termination and apoptosis.
Catalog No. NC9866266 Supplier Invivogen Supplier No. SUDGCV
Molecular Formula C9H13N5O4, Molecular weight 255.2, Ganciclovir (GCV) is a guanosine analog used as a prodrug to obtain a suicide effect in cells transfected with the herpes virus thymidine kinase gene (HSV-tk). HSVTK phosphorylates GCV to GCV-monophosphate which is further converted to GCV-diphosphate and GCV-triphosphate by host kinases. GCV-triphosphate causes premature DNA chain termination and apoptosis.