Learn More
Selleck Chemical LLC Ganetespib (STA-9090) S1159-50mg
SDP
Catalog No. 503112991
Encompass_Preferred
Ganetespib (STA-9090) is an HSP90 inhibitor with IC50 of 4 nM in OSA 8 cells induces apoptosis of OSA cells while normal osteoblasts are not affected active metabolite of STA-1474 Phase 3
Catalog No. 50-311-2991 Supplier Selleck Chemical LLC Supplier No. S115950MG
Ganetespib (STA-9090) is an HSP90 inhibitor with IC50 of 4 nM in OSA 8 cells induces apoptosis of OSA cells while normal osteoblasts are not affected active metabolite of STA-1474 Phase 3